Indium ≥99.9999%, shot
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-4943-25G
93-4943-5G
100207-296EA
366.58
USD
100207-296
100207-852
Indium ≥99.9999%, shot
Indium
Formula:
In MW: 114.82 g/mol Boiling Pt: 2000 Melting Pt: 156 Density: 7.31 |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...