Nickel(II) oxalate dihydrate

Supplier: STREM CHEMICALS INC MS

Synonyms: Nickel oxalate dihydrate

93-2831-50G 93-2831-250G
100211-246EA 200.98 USD
100211-246 100207-214
Nickel(II) oxalate dihydrate
Nickel(II) oxalate dihydrate
Formula: C₂H₄NiO₆
MDL Number: MFCD00167155
CAS Number: 6018-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR