Gold ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS

93-7912-500MG 93-7912-2G
100207-176EA 184 USD
100207-176 100206-060
Gold ≥99.9%, powder
Gold
Formula: Au
Boiling Pt: 2067 °C
Melting Pt: 1064 °C
Density: 19.3
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR