Tungsten disulfide ≥99.5% (W basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Tungsten sulfide, Tungsten sulphide, Tungsten (IV) sulfide, Tungsten (IV) sulphide
93-7424-50G
93-7424-250G
100199-662EA
114.65
USD
100199-662
100207-158
Tungsten disulfide ≥99.5% (W basis)
Tungsten disulphide
Formula:
S₂W Melting Pt: 1250 °C (dec.) Density: 7,5 |
MDL Number:
MFCD00011467 CAS Number: 12138-09-9 EINECS: 235-243-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...