Potassium nitrate ≥99% ACS

Supplier: STREM CHEMICALS INC MS
Warning

93-1956-500G 93-1956-2000G
100206-952EA 171.26 USD
100206-952 100200-064
Potassium nitrate ≥99% ACS
Potassium nitrate
Formula: KNO₃
Boiling Pt: 400 °C (dec.)
Melting Pt: 334 °C
Density: 2.109
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 13,07733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR