Potassium thiocyanate ≥99% ACS

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Potassium rhodanide

93-1952-250G 93-1952-50G
100205-392EA 101.91 USD
100205-392 100206-950
Potassium thiocyanate ≥99% ACS
Potassium thiocyanate
Formula: KSCN
Boiling Pt: 500 °C (1013 hPa)
Melting Pt: 175 °C
Density: 1.61 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR