Magnesium hydroxide ≥95%

Supplier: STREM CHEMICALS INC MS

93-1229-2000G
100206-920EA 40 USD
100206-920 100206-918
Magnesium hydroxide ≥95%
Magnesium hydroxide
Formula: Mg(OH)₂
MW: 58.32 g/mol
Density: 2.36
MDL Number: MFCD00011104
CAS Number: 1309-42-8
EINECS: 215-170-3
Merck Index: 13,05693

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR