Tin (II) trifluoromethanesulfonate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tin trifluoromethanesulfonate, Tin(II) trifluoromethanesulfonate, Tin trifluoromethanesulphonate, Tin triflate, Tin(II) trifluoromethanesulphonate

50-2750-10G 50-2750-2G
100206-748EA 387.82 USD
100206-748 100206-750
Tin (II) trifluoromethanesulfonate ≥99%
Tin (II) trifluoromethanesulphonate

Metal Triflate

Formula: C₂F₆O₆S₂Sn
Melting Pt: Dec.
MDL Number: MFCD00191251
CAS Number: 62086-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR