[P,P'-1,3-Bis(di-i-propylphosphino)propane][P-1,3-bis(di-i-propylphosphino)propane]palladium(0) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

46-0205-250MG 46-0205-1G
100198-816EA 420.37 USD
100198-816 100206-698
[P,P'-1,3-Bis(di-i-propylphosphino)propane][P-1,3-bis(di-i-propylphosphino)propane]palladium(0) ≥98%
[P,P'-1,3-Bis(di-i-propylphosphino)propane][P-1,3-bis(di-i-propylphosphino)propane]palladium(0)
MDL Number: MFCD06658138
CAS Number: 123333-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR