Allylchloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]palladium(II) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Allyl[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]chloropalladium(II), (NHC)Pd(allyl)Cl, Allylchloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]palladium(II), CX 21

46-0040-1G 46-0040-250MG
100206-694EA 359.51 USD
100206-694 100196-260
Allylchloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]palladium(II) ≥98%
Allylchloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]palladium(II)

NHC

Formula: C₃₀H₄₁ClN₂Pd
Melting Pt: 190 °C (dec.)
MDL Number: MFCD08561131
CAS Number: 478980-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR