Trimethyl(pentamethylcyclopentadienyl)titanium(IV) ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

22-5500-5G 22-5500-250MG 22-5500-1G
100206-484EA 1435.2 USD
100206-484 100198-706 100206-482
Trimethyl(pentamethylcyclopentadienyl)titanium(IV) ≥97%
Trimethyl(pentamethylcyclopentadienyl)titanium(IV)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₃H₂₄Ti
MDL Number: MFCD00269851
CAS Number: 107333-47-1
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR