2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2,8,9-Tris(2-methylpropyl)-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane, Triisobutylphosphatrane, 2,8,9-Tri-i-butyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane, [Nitrilotrisethylenetris(isobutylimino)]phosphine

15-6020-5G 15-6020-1G 15-6020-250MG
100206-408EA 669.48 USD
100206-408 100206-406 100198-668
2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane ≥97%
2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane

Phosphine

Formula: C₁₈H₃₉N₄P
MW: 342.51 g/mol
Density: 0.964
MDL Number: MFCD03840347
CAS Number: 331465-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR