Pentafluorobenzyl bromide ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: α-Bromo-2,3,4,5,6-pentafluorotoluene, 2,3,4,5,6-Pentafluorobenzyl bromide

09-5550-25G 09-5550-1G 09-5550-5G
100199-744EA 447.26 USD
100199-744 100206-270 100206-272
Pentafluorobenzyl bromide ≥98%
Pentafluorobenzyl bromide

Organo Fluorine

Formula: (C₆F₅)CH₂Br
Boiling Pt: 174…175 °C
Density: 1.728
Flash Pt: 181 °F
Storage Temperature: Ambient
MDL Number: MFCD00000299
CAS Number: 1765-40-8
EINECS: 217-182-4
UN: 1760
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR