Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cerium(IV) ≥97% (99.9%-Ce)

Supplier: STREM CHEMICALS INC MS

Synonyms: Ce(TMHD)4, Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)cerium(IV)

58-5000-25G 58-5000-5G 58-5000-1G
100206-122EA 2464.18 USD
100206-122 100206-774 100201-712
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cerium(IV) ≥97% (99.9%-Ce)
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cerium(IV)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₄₄H₇₆CeO₈
Boiling Pt: 295 °C/ 140 °C (0.05 mmHg) (dec./ subl.)
Melting Pt: 275…280 °C
MDL Number: MFCD00145378
CAS Number: 18960-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR