Cesium ≥99.9%

Supplier: STREM CHEMICALS INC MS
Danger


Prescored ampoule.

55-5551-1G 55-5551-5G 55-5551-25G
100209-370EA 242.03 USD
100209-370 100210-654 100206-114
Cesium ≥99.9%
Cesium
Formula: Cs
Boiling Pt: 690 °C
Melting Pt: 28.6 °C
Density: 1.892
MDL Number: MFCD00134037
CAS Number: 7440-46-2
EINECS: 231-155-4
UN: 1407
ADR: 4.3,I
Merck Index: 12,02051

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR