4-Carboxyphenylboronic acid ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 4-Boronobenzoic Acid, Ark Pharm, 4-Carboxybenzeneboronic acid

05-0400-1G 05-0400-5G
100201-306EA 56.62 USD
100201-306 100205-910
4-Carboxyphenylboronic acid ≥97%
4-Carboxyphenylboronic acid

Boronic Acid and Ester

Formula: C₇H₇BO₄
Melting Pt: 238…240 °C
MDL Number: MFCD00151801
CAS Number: 14047-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR