Tin(II) oxide ≥98% (99+%-Sn)

Supplier: STREM CHEMICALS INC MS

93-5039-50G 93-5039-250G
100210-440EA 63.69 USD
100210-440 100205-886
Tin(II) oxide ≥98% (99+%-Sn)
Tin (II) oxide
Formula: OSn
Boiling Pt: 1425 °C (1013 hPa)
Melting Pt: Dec.
Density: 6.95
MDL Number: MFCD00011243
CAS Number: 21651-19-4
EINECS: 244-499-5
Merck Index: 12,08944

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR