Mercury(II) acetate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Mercury diacetate, Mercuric acetate, Acetic acid mercury (II) salt

93-8040-25G 93-8040-100G
100205-780EA 89.17 USD
100205-780 100210-570
Mercury(II) acetate ≥98%
Mercury (II) acetate
Formula: (H₃CCOO)₂Hg
Melting Pt: Dec.
Density: 3.270
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II
Merck Index: 13,05898

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR