Holmium ≥99.9%, ingot

Supplier: STREM CHEMICALS INC MS

93-6732-5G 93-6732-25G
100205-736EA 229.29 USD
100205-736 100202-574
Holmium ≥99.9%, ingot
Holmium
Formula: Ho
Boiling Pt: 2695 °C
Melting Pt: 1474 °C
Density: 8,795
MDL Number: MFCD00011049
CAS Number: 7440-60-0
EINECS: 231-169-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR