Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')europium(III) ≥99% (99.9%-Eu)

Supplier: STREM CHEMICALS INC MS

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III), Eu(TMHD)3

93-6328-5G 93-6328-1G
100201-284EA 304.31 USD
100201-284 100205-720
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')europium(III) ≥99% (99.9%-Eu)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')europium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₃H₅₇EuO₆
Boiling Pt: 275 °C (dec.)
Melting Pt: 188…189 °C
MDL Number: MFCD00000019
CAS Number: 15522-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR