Lanthanum(III) fluoride, anhydrous ≥99.9% (La basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Lanthanum trifluoride

93-5710-100G 93-5710-25G
100210-488EA 256.18 USD
100210-488 100205-670
Lanthanum(III) fluoride, anhydrous ≥99.9% (La basis)
Lanthanum (III) fluoride
Formula: F₃La
Boiling Pt: ∼2327 °C (1013 hPa)
Melting Pt: 1493 °C
Density: 5.936
Storage Temperature: Ambient
MDL Number: MFCD00011073
CAS Number: 13709-38-1
EINECS: 237-252-8
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR