Silver tetrafluoroborate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Argentous tetrafluoroborate

93-4734-25G 93-4734-5G
100205-592EA 281.66 USD
100205-592 100209-120
Silver tetrafluoroborate ≥99%
Silver tetrafluoroborate
Formula: AgBF₄
Melting Pt: 200 °C (dec.)
MDL Number: MFCD00003408
CAS Number: 14104-20-2
EINECS: 237-956-5
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR