Molybdenum(IV) sulfide ≥97%

Supplier: STREM CHEMICALS INC MS

Synonyms: Molybdenum disulfide, Molybdenum disulphide

93-4247-500G 93-4247-100G
100208-178EA 403.38 USD
100208-178 100205-578
Molybdenum(IV) sulfide ≥97%
Molybdenum (IV) sulphide
Formula: MoS₂
Melting Pt: 1185 °C
Density: 4.80
MDL Number: MFCD00003470
CAS Number: 1317-33-5
EINECS: 215-263-9
Merck Index: 12,06318

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR