Iron(III) nitrate nonahydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate

93-2614-5000G 93-2614-250G 93-2614-1000G
100207-808EA 172.68 USD
100207-808 100205-456 100200-084
Iron(III) nitrate nonahydrate ≥98%
Iron (III) nitrate nonahydrate
Formula: Fe(NO₃)₃·9H₂O
MW: 404 g/mol
Boiling Pt: 125 (dec.)
Melting Pt: 47
Density: 1.684
Storage Temperature: Ambient
MDL Number: MFCD00149708
CAS Number: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1 III
Merck Index: 13,04057

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR