Manganese ≥99.5%, powder

Supplier: STREM CHEMICALS INC MS
Warning

93-2543-1000G 93-2543-250G
100210-286EA 309.97 USD
100210-286 100205-454
Manganese ≥99.5%, powder
Manganese
Formula: Mn
Boiling Pt: 1900 °C
Melting Pt: 1260 °C
Density: 7.20
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.1,III
Merck Index: 12,05762

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR