Vanadium(V) oxide ≥99.5%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Vanadium pentoxide, di-Vanadium pentaoxide
93-2321-500G
93-2321-100G
100205-430EA
257.6
USD
100205-430
100208-528
Vanadium(V) oxide ≥99.5%
Vanadium (V) oxide
Formula:
O₅V₂ Boiling Pt: 1750 °C (dec.) Melting Pt: 690 °C Density: 3.357 |
MDL Number:
MFCD00011457 CAS Number: 1314-62-1 EINECS: 215-239-8 UN: 2862 ADR: 6.1,III Merck Index: 13,09987 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...