Titanium ≥99.5%, rods

Supplier: STREM CHEMICALS INC MS
Warning

93-2270-10CM 93-2270-50CM
100205-426EA 70.77 USD
100205-426 100199-518
Titanium ≥99.5%, rods
Titanium
Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.5
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR