Magnesium sulfate heptahydrate ≥98% ACS

Supplier: STREM CHEMICALS INC MS

Synonyms: sulphuric acid magnesium salt heptahydrate, sulfuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

93-1256-250G 93-1256-1000G
100205-298EA 42.46 USD
100205-298 100204-162
Magnesium sulfate heptahydrate ≥98% ACS
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
Density: 1.68
MDL Number: MFCD00149785
CAS Number: 10034-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR