Dichlorodicarbonylbis(triphenylphosphine)ruthenium(II) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

44-0400-1G 44-0400-250MG
100200-684EA 386.4 USD
100200-684 100205-126
Dichlorodicarbonylbis(triphenylphosphine)ruthenium(II) ≥98%
Dichlorodicarbonylbis(triphenylphosphine)ruthenium(II)
Formula: C₃₈H₃₄Cl₂O₂P₂Ru
MDL Number: MFCD00009594
CAS Number: 14564-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR