Potassium fluoride, anhydrous ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

93-1922-25G 93-1922-100G 93-1922-500G
100197-722EA 36.8 USD
100197-722 100201-944 100204-682
Potassium fluoride, anhydrous ≥98%
Potassium fluoride
Formula: KF
Boiling Pt: 1505 °C
Melting Pt: 846 °C
Density: 2.48
Storage Temperature: Ambient
MDL Number: MFCD00011398
CAS Number: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III
Merck Index: 13,07714

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR