Sulfur ≥99%, powder, sublimated

Supplier: STREM CHEMICALS INC MS
Warning

93-1618-250G 93-1618-1000G
100204-196EA 50.95 USD
100204-196 100204-676
Sulfur ≥99%, powder, sublimated
Sulphur
Formula: S
Boiling Pt: 444.67 °C
Melting Pt: 112.8 °C
Density: 2.07
Flash Pt: 405 °F
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1,III
Merck Index: 13,09059

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR