Beryllium ≥99%, powder

Supplier: STREM CHEMICALS INC MS
Danger

93-0420-5G 93-0420-25G
100201-814EA 72.18 USD
100201-814 100204-664
Beryllium ≥99%, powder
Beryllium
Formula: Be
Boiling Pt: 2970 °C (5 mmHg)
Melting Pt: 1278 °C
Density: 1,85
MDL Number: MFCD00134032
CAS Number: 7440-41-7
EINECS: 231-150-7
UN: 1567
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR