Bismuth ≥99.9999%, granules
Supplier: STREM CHEMICALS INC MS
83-0050-250G
83-0050-50G
83-0050-10G
100204-658EA
2563.26
USD
100204-658
100203-336
100208-976
Bismuth ≥99.9999%, granules
Bismuth
Formula:
Bi Boiling Pt: 1560 °C Melting Pt: 271.3 °C Density: 9.80 |
MDL Number:
MFCD00134033 CAS Number: 7440-69-9 EINECS: 231-177-4 Merck Index: 12,01297 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...