Osmium tetraoxide ≥99.95% (Os basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid

76-2950-5G 76-2950-500MG 76-2950-1G 76-2950-250MG
100204-648EA 833.66 USD
100204-648 100199-448 100202-688 100201-736
Osmium tetraoxide ≥99.95% (Os basis)
Osmium tetraoxide
Formula: O₄Os
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: 41…42 °C
Density: 4.906
Storage Temperature: Ambient
MDL Number: MFCD00011150
CAS Number: 20816-12-0
EINECS: 244-058-7
UN: 2471
ADR: 6.1,I
Merck Index: 13,06962

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR