Nickel (II) oxide ≥99.998% (Ni basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

28-1475-50G 28-1475-10G
100206-104EA 515.2 USD
100206-104 100204-608
Nickel (II) oxide ≥99.998% (Ni basis), PURATREM
Nickel (II) oxide
Formula: NiO
Melting Pt: 1990 °C
Density: 6.67
MDL Number: MFCD00011145
CAS Number: 1313-99-1
EINECS: 215-215-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR