(S)-(+)-1-[(R)-2-(Di-2-furylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (2S)-1-[(1S)-1-[Bis(3,5-dimethylphenyl)phosphino]ethyl]-2-(di-2-furanylphosphino)ferrocene

26-1170-100MG 26-1170-500MG
100209-832EA 157.11 USD
100209-832 100204-580
(S)-(+)-1-[(R)-2-(Di-2-furylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine ≥97%
(S)-(+)-1-[(R)-2-(Di-2-furylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine

JOSIPHOS, Phosphine

Formula: C₃₆H₃₆FeO₂P₂
MDL Number: MFCD06658128
CAS Number: 649559-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR