Diethylphosphine ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

15-1210-1G 15-1210-5G
100204-510EA 236.37 USD
100204-510 100204-512
Diethylphosphine ≥99%
Diethylphosphine

Phosphine

Boiling Pt: 85 °C
Density: 0,7862
MDL Number: MFCD00015169
CAS Number: 627-49-6
UN: 2845
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR