Tungsten(IV) carbide ≥99.5% (W basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Tungsten monocarbide, Wolfram carbide, Tungsten carbide

93-7431-50G 93-7431-250G
100204-424EA 55.2 USD
100204-424 100205-760
Tungsten(IV) carbide ≥99.5% (W basis)
Tungsten(IV) carbide
Formula: CW
Boiling Pt: 6000 °C
Melting Pt: ~2870 °C
Density: 15.63
MDL Number: MFCD00011464
CAS Number: 12070-12-1
EINECS: 235-123-0
UN: 3178
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR