Gadolinium ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS

93-6433-50X50MM 93-6433-25X25MM
100201-286EA 925.66 USD
100201-286 100204-408
Gadolinium ≥99.9%, foil
Gadolinium
Formula: Gd
MW: 157.25
Boiling Pt: 3233 °C
Melting Pt: 1311 °C
Density: 7.898
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR