Lanthanum triacetate hydrate ≥99.9% (La basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-5701-100G 93-5701-500G
100204-380EA 94.83 USD
100204-380 100200-586
Lanthanum triacetate hydrate ≥99.9% (La basis)
Lanthanum triacetate hydrate
Formula: C₆H₉LaO₆·H₂O
MDL Number: MFCD00150118
CAS Number: 100587-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR