Cesium sulfate ≥99.9% (Cs basis)
Supplier: STREM CHEMICALS INC MS
93-5547-50G
93-5547-10G
100205-658EA
182.58
USD
100205-658
100204-366
Cesium sulfate ≥99.9% (Cs basis)
Cesium sulphate
Formula:
Cs₂O₄S Boiling Pt: 1627 °C (1013 hPa) Melting Pt: 1010 °C Density: 4.243 Storage Temperature: Ambient |
MDL Number:
MFCD00010959 CAS Number: 10294-54-9 EINECS: 233-662-6 Merck Index: 13,02025 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...