Calcium nitrate tetrahydrate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-2028-250G
93-2028-1000G
100204-224EA
55.2
USD
100204-224
100204-222
Calcium nitrate tetrahydrate ≥99%
Calcium nitrate tetrahydrate
Formula:
Ca(NO₃)₂·4H₂O Boiling Pt: 132 °C (dec.) Melting Pt: ∼42 °C Density: 1.82 Storage Temperature: Ambient |
MDL Number:
MFCD00149604 CAS Number: 13477-34-4 EINECS: 233-332-1 UN: 1454 ADR: 5.1,III Merck Index: 13,01687 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...