Calcium nitrate tetrahydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-2028-250G 93-2028-1000G
100204-224EA 55.2 USD
100204-224 100204-222
Calcium nitrate tetrahydrate ≥99%
Calcium nitrate tetrahydrate
Formula: Ca(NO₃)₂·4H₂O
Boiling Pt: 132 °C (dec.)
Melting Pt: ∼42 °C
Density: 1.82
Storage Temperature: Ambient
MDL Number: MFCD00149604
CAS Number: 13477-34-4
EINECS: 233-332-1
UN: 1454
ADR: 5.1,III
Merck Index: 13,01687

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR