Magnesium sulfate heptahydrate ≥98% ACS
Supplier: STREM CHEMICALS INC MS
Synonyms:
sulphuric acid magnesium salt heptahydrate, sulfuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt
93-1256-250G
93-1256-1000G
100205-298EA
42.46
USD
100205-298
100204-162
Magnesium sulfate heptahydrate ≥98% ACS
Magnesium sulphate heptahydrate
Formula:
MgSO₄·7H₂O Density: 1.68 |
MDL Number:
MFCD00149785 CAS Number: 10034-99-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...