Cobalt octoate ∼65% in mineral spirits (12% Co)

Supplier: STREM CHEMICALS INC MS
Danger

27-0449-250G 27-0449-1000G 27-0449-50G
100204-020EA 104.74 USD
100204-020 100209-876 100209-878
Cobalt octoate ∼65% in mineral spirits (12% Co)
Formula: C₁₆H₃₀CoO₄
Density: 1.01
Flash Pt: 104 °F
MDL Number: MFCD00072632
CAS Number: 136-52-7
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR