Vanadium(III) chloride, anhydrous ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
23-4300-50G
23-4300-10G
100204-012EA
301.48
USD
100204-012
100204-010
Vanadium(III) chloride, anhydrous ≥95%
Vanadium (III) chloride
Formula:
Cl₃V Density: 3.00 |
MDL Number:
MFCD00011454 CAS Number: 7718-98-1 EINECS: 231-744-6 UN: 2475 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...