Tetramethylphosphonium bromide ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-5360-5G 15-5360-1G
100203-988EA 549.17 USD
100203-988 100203-986
Tetramethylphosphonium bromide ≥98%
Tetramethylphosphonium bromide
Formula: C₄H₁₂BrP
MDL Number: MFCD00011802
CAS Number: 4519-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR