Ferrocenecarboxylic acid ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ferrocene monocarboxylic acid

26-1900-1G 26-1900-25G 26-1900-5G
100197-468EA 66.52 USD
100197-468 100207-246 100203-760
Ferrocenecarboxylic acid ≥97%
Ferrocenecarboxylic acid
Formula: C₁₁H₁₀FeO₂
Melting Pt: 210 °C (dec.)
MDL Number: MFCD00001430
CAS Number: 1271-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR