Hafnium dioxide ≥99.995% (Hf basis) (<0.15% Zr), PURATREM

Supplier: STREM CHEMICALS INC MS

72-5200-5G 72-5200-1G
100203-676EA 611.45 USD
100203-676 100209-376
Hafnium dioxide ≥99.995% (Hf basis) (<0.15% Zr), PURATREM
Hafnium dioxide
Formula: HfO₂
Boiling Pt: 5400 °C (1013 hPa)
Melting Pt: 2812 °C
Density: 9.68
MDL Number: MFCD00003565
CAS Number: 12055-23-1
EINECS: 235-013-2
Merck Index: 14,04727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR