Cerium trinitrate hexahydrate ≥99.99% (Ce basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cerous chloride hexahydrate, Cerium (III) nitrate hexahydrate

93-5814-50G 93-5814-10G
100206-048EA 304.31 USD
100206-048 100203-528
Cerium trinitrate hexahydrate ≥99.99% (Ce basis), PURATREM
Cerium trinitrate hexahydrate
Formula: Ce(NO₃)₃·6H₂O
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 39 °C
Density: 2.377 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149631
CAS Number: 10294-41-4
EINECS: 233-297-2
UN: 1477
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR